Learn More
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Product Code. 11581851
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Irradiated, 25 to 40 doses
None
Unit Size:
1 L
1000mL
100mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
11581851 500 mL None 500mL
11571851 100 mL None 100mL
11556551 1000 mL None 1000mL
16291102 500 mL Irradiated, 25 to 40 doses 500mL
16605755 1000 mL Irradiated, 25 to 40 doses 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11581851 Supplier Cytiva Supplier No. SH30118.03
This item is not returnable. View return policy
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
  • Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
TRUSTED_SUSTAINABILITY
Specifications
Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Description Newborn Calf Serum (U.S.)
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus