Learn More
Cytiva HyClone™ Cell Boost Kit
Product Code. 10237053
Change view
Click to view available options
Quantity:
100 g
Unit Size:
6g
Product Code. Quantity Unit Size
10237053 100 g 6g
1 options
This item is not returnable. View return policy
Product Code. 10237053 Supplier Cytiva Supplier No. SH30890
 Restricted Item
Fecha estimada de envoi: 08-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Content And Storage 2° to 8° C
Description Cell Boost Kit (1 through 6)
Form Powder
Quantity 100 g