Learn More
Cytiva HyClone™ Cell Boost™ 6 Supplement
Product Code. 11576341
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
11576341 100 g 100g
1 options
This item is not returnable. View return policy
Product Code. 11576341 Supplier Cytiva Supplier No. SH30866.01
  • ONLINE EXCLUSIVE PRICE
    327.54€
    288.00€ / 100 g
    Ahorro 39.54€
    12% Descuento
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Description Cell Boost 6 (CN-T)
Classification Chemically-defined, Protein-free, Animal Origin-free
Quantity 100 g