Learn More
Human TCF7 knockout HeLa cell lysate, 1 Kit
Product Code. 32143138 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
32143138 Each
1 options
This item is not returnable. View return policy
Product Code. 32143138 Supplier Abcam Supplier No. ab2582251Kit
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Human TCF7 knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: TCF7 knockout HeLa
Specifications
For Use With (Application) Sanger seq