Learn More
Human PARP10 (ARTD10) knockout A549 cell lysate, 1 Kit
Product Code. 32157402 Shop All Abcam Products
This item is not returnable. View return policy
Product Code. 32157402 Supplier Abcam Supplier No. ab2585641Kit
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Human PARP10 (ARTD10) knockout A549 cell/tissue lysate.
  • Species: Human
  • Tissue: PARP10 (ARTD10) knockout A549