Learn More
Human NEIL1 knockout HEK-293T cell lysate, 1 Kit
Product Code. 32154322 Shop All Abcam Products
This item is not returnable. View return policy
Product Code. 32154322 Supplier Abcam Supplier No. ab2589961Kit
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Human NEIL1 knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: NEIL1 knockout HEK-293T