Learn More
Human MYC (c-Myc) knockout HEK-293T cell lysate, 1 Kit
Product Code. 32142006 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
32142006 Each
1 options
This item is not returnable. View return policy
Product Code. 32142006 Supplier Abcam Supplier No. ab2638501Kit
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Human MYC (c-Myc) knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: MYC (c-Myc) knockout HEK-293T
Specifications
For Use With (Application) Western Blot