Learn More
Human BATF2 (SARI) knockout A549 cell lysate, 1 Kit
Product Code. 32156299 Shop All Abcam Products
This item is not returnable. View return policy
Product Code. 32156299 Supplier Abcam Supplier No. ab2578541Kit
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Human BATF2 (SARI) knockout A549 cell/tissue lysate.
  • Species: Human
  • Tissue: BATF2 (SARI) knockout A549