Learn More
Hexamethylacetone, 98%
Description
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 0.824 |
| Boiling Point | 152°C to 153°C |
| Flash Point | 34°C (93°F) |
| Refractive Index | 1.419 |
| Quantity | 25 g |
| UN Number | UN1224 |
| Beilstein | 1701122 |
| Synonym | hexamethylacetone, 2,2,4,4-tetramethyl-3-pentanone, di-tert-butyl ketone, 3-pentanone, 2,2,4,4-tetramethyl, 2,2,4,4-tetramethyl-pentan-3-one, 3-pentanone,2,2,4,4-tetramethyl, di-t-butyl ketone, acmc-20akrk, t-c4h9 2co, 2,4,4-tetramethyl-3-pentanone |
| Formula Weight | 142.24 |
| Percent Purity | 98% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.