Learn More
Lonza™ hEGF Epidermal Growth Factor
Product Code. 11625490
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
11625490 0.5 mL 1mL
1 options
This item is not returnable. View return policy
Product Code. 11625490 Supplier Lonza™ Supplier No. CC4107
  • ONLINE EXCLUSIVE PRICE
    424.00€
    331.47€ / 1 ml
    Ahorro 92.53€
    21% Descuento
 Restricted Item
Fecha estimada de envoi: 15-12-2026