Learn More
Glutathione oxidized, MedChemExpress
Product Code. 16408916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
16408916 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 16408916 Supplier Medchem Express Supplier No. HYD0844
 Restricted Item
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Glutathione oxidized (L-Glutathione oxidized) is produced by the oxidation of glutathione which is a major intracellular antioxidant and detoxifying agent.
Glutathione oxidized (L-Glutathione oxidized) is produced by the oxidation of glutathione which is a major intracellular antioxidant and detoxifying agent.
Specifications
Chemical Name or Material Glutathione oxidized
CAS 27025-41-8
Color White
Purity Grade Notes Research
Molecular Formula C20H32N6O12S2
Quantity 100 mg
Synonym L-Glutathione oxidized GSSG Oxiglutatione
Solubility Information H2O : 250 mg/mL (408.08 mM; Need ultrasonic)
SMILES O=C(NCC(O)=O)[C@H](CSSC[C@@H](C(NCC(O)=O)=O)NC(CC[C@H](N)C(O)=O)=O)NC(CC[C@H](N)C(O)=O)=O
Molecular Weight (g/mol) 612.63
Formula Weight 612.63
Percent Purity 98.07%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism