Learn More
Glutamate Receptor 6 Antibody (8A1F11) - BSA Free, R&D Systems™
Product Code. 18666768 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18666768 0.025 mL 0.03mL
18283421 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18666768 Supplier Novus Biologicals Supplier No. NBP2617880.025ml
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Glutamate Receptor 6 Monoclonal specifically detects Glutamate Receptor 6 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Glutamate Receptor 6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 8A1F11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427
Gene Symbols GRIK2
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIK2 (AA: extra 45-226) expressed in E. Coli.
Molecular Weight of Antigen 102.5 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2898
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus