Includes
Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.
- Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
- No need to remove unincorporated nucleotides before hybridization
- Protocols for labeling directly from low melting point agarose
Specifications
Specifications
| Content And Storage | -15°C to -30°C |
| For Use With (Application) | dNTP Labeling |
| Includes | Primer solution 150μL; Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 Nucleotide solutions: dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA 120μL, Reaction Buffer A 10 x concentra |
| No. of Reactions | 30 |
| Product Type | Megaprime DNA Labeling System, dNTP (for use with any radioactively labeled nucleotide) |
| Quantity | 30 reactions |