Learn More
R&D Systems™ GAPDH Primer
Product Code. 18748403 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
Product Code. Quantity Unit Size
18748403 0.5 mL 0.5mL
1 options
This item is not returnable. View return policy
Product Code. 18748403 Supplier R&D Systems™ Supplier No. NBP171650
  • ONLINE EXCLUSIVE PRICE
    170.01€
    144.90€ / 0.5 ml
    Ahorro 25.11€
    14% Descuento
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Novus
Quantity 0.5 mL
For Use With (Application) ChIP