Learn More
Aalborg™ PWE Paddlewheel Meter
Product Code. 11728628
Change view
Click to view available options
Flow Rate Range:
0.3 to 38 LPM
0.6 to 64 LPM
Unit Size:
Each
Product Code. Flow Rate Range unitSize
11728628 0.6 to 64 LPM Each
11708628 0.3 to 38 LPM Each
2 options
This item is not returnable. View return policy
Product Code. 11728628
Brand: Aalborg™ PWE08PELRB2
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Aalborg™ PWE Paddlewheel Meter measures the flow of liquids by allowing the liquid to flow through the unit, causing the paddle wheel to spin.
  • Supports up to 23 Engineering Units (including one User Defined)
  • Two programmable Totalizers (main and pilot) indicate total liquid volume
  • Programmable High and low Flow Alarm limits with preset delay interval
  • Programmable High and low Temperature Alarm limits with preset delay interval (optional)
  • Two sets of user programmable optically isolated outputs
  • User selectable (via jumpers) analog 0-5 Vdc or 4-20mA flow and temperature (optional) outputs
  • Flow Pulse output (3.3Vdc CIMOS)
  • Digital Interface (RS-232 or RS-485)
  • Multi-Drop Capability of up to 256 units (RS-485 option)
  • Extensive Self Diagnostic with LCD (optional) indication
  • Local key pad and 2x16 characters LCD display with adjustable back light (optional)
  • Enclosure weather tight to IP65 standards
Specifications
Type Flowmeter
Accuracy ±1%
Pressure Rating 150 psig
Flow Rate Range 0.6 to 64 LPM
Repeatability ±0.25% (Full Scale)
Temperature (Metric) -10°C to 70°C
Viscosity Calibrated to 1 cSt (Water)