Learn More
Gibco™ Expi293™ MembranePro™ Expression System
Expi293™ MembranePro™ Expression System
Especificaciones
| Tipo | Sistema de expresión |
|---|---|
| Tipo de entrega | Partículas similares a virus (VLP) |
| Línea de células | Expi293F™ |
| Para utilizar con (aplicación) | Expresión de proteínas de membrana |
| Promotor | EF-1a |
Descripción
For researchers studying G protein–coupled receptors (GPCRs) and other membrane proteins, the Expi293™ MembranePro™ Expression System delivers enriched, functional membrane proteins with high efficiency, scalability and reliability for a wide variety of low or high throughput applications.Features of the Expi293™ MembranePro™ Expression System include:
In addition, benefits of MembranePro™ technology include:
With the less labor-intensive Expi293™ MembranePro™ Expression System protocol, you will:
1. Clone your gene of interest using a TOPO™ vector
2. Transfect a susupension culture of Expi293F™ cells using Expifectamine™ 293 transfection reagent
3. Harvest lipoparticles 48 hrs after addition of enhancers using MembranePro™ Precipitation Mix
4. Store lipoparticles at -80 C or proceed to downstream assays
Note: Exi293F™ Cells (A14527) and a pEF6 V5-His TOPO™ Expression Kit (K9610-20) are required for use of this system and must be purchased separately when needed.
Especificaciones
| Sistema de expresión | |
| Expi293F™ | |
| EF-1a |
| Partículas similares a virus (VLP) | |
| Expresión de proteínas de membrana |
For Research Use Only. Not for use in diagnostic procedures.