Learn More
ETV6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16801274 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16801274 20 μL 20µL
16891264 150 μL 150µL
2 options
This item is not returnable. View return policy
Product Code. 16801274
Brand: Proteintech 131821AP20UL
This item is not returnable. View return policy
Rabbit Polyclonal Antibody
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
Specifications
Antigen ETV6
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ETV6
Gene Accession No. P41212, P97360
Gene Alias ETS related protein Tel1, ETS translocation variant 6, ets variant 6, ETV6, TEL, TEL/ABL, TEL1, Transcription factor ETV6
Gene Symbols Etv6
Host Species Rabbit
Immunogen ETV6 Fusion Protein Ag3852
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14011, 2120, 312777
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG