Learn More
MilliporeSigma™ Silica Gel 60 F254 Coated Glass-Backed TLC Plates
Allows analyzation of any substance when combined with suitable mobile phase.
Description
- Combination of unmodified silica gel 60 and polymeric binder for hard and adherent surface, resistant to crack and damage
- Dense plate coating ensures narrow bands and maximum sample separation efficiency
- Layer thickness of 250μm with mean particle size of 10 to 12μm
- Green fluorescencing inorganic florescent indications for UV detection of colorless substance
- Disposable
- Available in broad range of sizes
- Fibre case, paper box and plastic box packaging options
Applications:
- In-process control in drug synthesis
- Identity and stability testing of drugs
- Quality control of pharmaceuticals, food and environmental samples
- Residue analysis in food and environmental samples