Learn More
Ebrotidine, MedChemExpress
Product Code. 16486608
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486608 10 mg 10mg
16469177 5 mg 5mg
16402707 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486608 Supplier Medchem Express Supplier No. HY15538
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Ebrotidine(FI 3542) is a competitive H2-receptor antagonist (Ki= 127.5 nM) with a potent antisecretory activity and evidenced gastroprotection.
Ebrotidine(FI 3542) is a competitive H2-receptor antagonist (Ki= 127.5 nM) with a potent antisecretory activity and evidenced gastroprotection.
Specifications
Chemical Name or Material Ebrotidine
CAS 100981-43-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H17BrN6O2S3
Quantity 10 mg
Synonym FI3542
Solubility Information DMSO : 100 mg/mL (209.46 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(Br)C=C1)(/N=C/NCCSCC2=CSC(NC(N)=N)=N2)=O
Molecular Weight (g/mol) 477.42
Formula Weight 477.42
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation