Learn More
STEMCELL Technologies EasySep™ Magnet
Product Code. 17167841
Encompass_Preferred
Change view
Click to view available options
Quantity:
2.5 x 108 Cells
Unit Size:
1 set
Product Code. Quantity Unit Size
17167841 2.5 x 108 Cells 1 set
1 options
This item is not returnable. View return policy
Product Code. 17167841 Supplier STEMCELL Technologies Supplier No. 18000
 Restricted Item
Fecha estimada de envoi: 18-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Immunomagnetic column-free magnet
  • The EasySep™ Magnet is designed for cell separation procedures using EasySep™ reagents
  • The EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ Magnetic Particles without the use of columns
  • This magnet is designed to hold a standard 12 x 75 mm (5 mL) polystyrene tube
Specifications
For Use With (Application) Cell Isolation
Quantity 2.5 x 108 Cells