Learn More
(E)-Cardamonin, MedChemExpress
Product Code. 16473916
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473916 25 mg 25mg
16461628 5 mg 5mg
16439156 100 mg 100mg
16429218 10 mg 10mg
16414917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473916 Supplier Medchem Express Supplier No. HYN1378
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress (E)-Cardamonin ((E)-Cardamomin) is a novel antagonist of hTRPA1 cation channel with an IC50 of 454 nM.
(E)-Cardamonin ((E)-Cardamomin) is a novel antagonist of hTRPA1 cation channel with an IC50 of 454 nM.
Specifications
Chemical Name or Material (E)-Cardamonin
CAS 19309-14-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H14O4
Quantity 25 mg
Synonym (E)-Cardamomin (E)-Alpinetin chalcone
Solubility Information DMSO : ≥ 28 mg/mL (103.60 mM)
SMILES O=C(C1=C(OC)C=C(O)C=C1O)/C=C/C2=CC=CC=C2.[(E)]
Molecular Weight (g/mol) 270.28
Formula Weight 270.28
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death