Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock, with short inlet, complete with SBW-J key
Product Code. 15864748 Shop All DWK Life Sciences Products
Change view
Click to view available options
Plug Bore:
2 mm
2.5 mm
4 mm
Unit Size:
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore unitSize
15864748 4 mm Pack of 25
10709021 2.5 mm Pack of 25
15854748 2 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15864748 Supplier DWK Life Sciences Supplier No. 286432405
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For easier attachment to DURAN™ separating funnel raw bodies.
DURAN™ Separating funnel taps with short funnels are components for the production of separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape up to the socket of the valve.
  • Separating funnel tap sleeve with SBW-J plug
  • with short approach funnel
  • With a particularly strong approach to reducing the risk of breakage
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 4 mm
Material Borosilicate Glass (3.3), SBW-J, Polypropylene
Autoclavable Not Autoclavable
Closure Size 18.8 NS
No. per Pack 25/Pk.
For Use With (Application) For attachment to DURAN™ separating funnel raw bodies