Learn More
Kartell™ Douche Can
Product Code. 10396251
Change view
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10396251 1000 mL Each
12621006 2000 mL Each
2 options
This item is not returnable. View return policy
Product Code. 10396251
Brand: Kartell™ 0058900
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Kartell™ Douche Can is a tool for irrigation and is provided with a falt back and suspension tag.
  • Provided with flat back
  • Includes suspension tag
  • Autoclavable
Specifications
Description Douche can or irrigator
Capacity (Metric) 1000 mL
Material Polypropylene
Shape Cylindrical
Type Douche Can
Diameter (Metric) 9.9 mm