Learn More
Diphenylmethylsilane, 95%, BLD Pharm™
Product Code. 31450069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31450069 5 g
31435267 25 g
31434926 1 g
31433890 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31450069 Supplier BLD Pharm Supplier No. BD1259105g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 776-76-1
Molecular Formula C13H14Si
MDL Number MFCD00008255
Quantity 5 g
SMILES C[SiH](C1=CC=CC=C1)C2=CC=CC=C2
Molecular Weight (g/mol) 198.34
Percent Purity 95%
Chemical Name or Material Diphenylmethylsilane