missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl carbonate, 99%

Product Code. 11419503
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
Product Code. Quantity unitSize
11419503 5000 g 5000g
11409503 250 g 250g
11499493 1000 g 1000g
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11419503

Brand: Thermo Scientific Alfa Aesar A12362.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 102-09-0 | C13H10O3 | 214.22 g/mol

Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-09-0
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00003037
InChI Key ROORDVPLFPIABK-UHFFFAOYSA-N
Synonym carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
PubChem CID 7597
ChEBI CHEBI:34722
IUPAC Name diphenyl carbonate
SMILES C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2

Specifications

Melting Point 76°C to 81°C
Density 1.3
Boiling Point 301°C to 302°C
Flash Point 168°C (334°F)
Odor Mild
Quantity 5000 g
Beilstein 1074863
Merck Index 14,3602
Solubility Information Soluble in ethanol,diethyl ether,carbon tetrachloride,acetic acid. Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material Diphenyl carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.