Learn More
Dioctyltin oxide, 98%, BLD Pharm™
Product Code. 31447972
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31447972 500 g
31442160 25 g
31443911 100 g
31437928 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31447972 Supplier BLD Pharm Supplier No. BD125733500g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 870-08-6
Molecular Formula C16H34OSn
MDL Number MFCD00013839
Quantity 500 g
SMILES CCCCCCCC[Sn](CCCCCCCC)=O
Molecular Weight (g/mol) 361.15
Percent Purity 98%
Chemical Name or Material Dioctyltin oxide