Learn More
Dihydrostreptomycin sulfate, BLD Pharm™
Specifications
Specifications
| CAS | 5490-27-7 |
| Molecular Formula | C42H88N14O36S3 |
| MDL Number | MFCD02094085 |
| Quantity | 25 g |
| SMILES | O[C@]1([C@H]([C@H](O[C@@H]2[C@H]([C@@H]([C@H]([C@@H]([C@H]2O)O)NC(N)=N)O)NC(N)=N)O[C@H]1C)O[C@H]3[C@H]([C@@H]([C@H]([C@@H](O3)CO)O)O)NC)CO.O[C@]4([C@H]([C@H](O[C@@H]5[C@H]([C@@H]([C@H]([C@@H]([C@H]5O)O)NC(N)=N)O)NC(N)=N)O[C@H]4C)O[C@H]6[C@H]([C@@H]([C@H]([C@@H](O6)CO)O)O)NC)CO.O=S(O)(O)=O.O=S(O)(O)=O.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 1461.42 |
| Chemical Name or Material | Dihydrostreptomycin sulfate |