Learn More
Diethyl isobutylmalonate, 98%, BLD Pharm™
Product Code. 31033234
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31033234 500 g
31032356 100 g
31029201 10 g
31024778 25 g
31027095 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31033234 Supplier BLD Pharm Supplier No. BD13487500g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Especificaciones
CAS 10203-58-4
Molecular Formula C11H20O4
MDL Number MFCD00026869
Quantity 500 g
SMILES O=C(OCC)C(CC(C)C)C(OCC)=O
Molecular Weight (g/mol) 216.27
Percent Purity 98%
Chemical Name or Material Diethyl isobutylmalonate