Learn More
Diethyl (2-oxo-2-phenylethyl)phosphonate, 98%, BLD Pharm™
Product Code. 30958954
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30958954 1 g -
30949117 5 g 5g
30952410 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30958954 Supplier BLD Pharm Supplier No. BD1245041g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3453-00-7
Molecular Formula C12H17O4P
MDL Number MFCD00015342
Quantity 1 g
SMILES O=C(C1=CC=CC=C1)CP(OCC)(OCC)=O
Molecular Weight (g/mol) 256.23
Percent Purity 98%
Chemical Name or Material Diethyl (2-oxo-2-phenylethyl)phosphonate