Learn More
DGK-delta Antibody - Azide and BSA Free, R&D Systems™
Produktkode 18613191 Shop alle Bio Techne produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.02 mL
0.1 mL
Förpackningsstorlek:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity Förpackningsstorlek
18613191 0.02 mL 0.02mL
18647220 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 18613191 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP2922070.02ml
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolitik
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
DGK-delta Polyclonal antibody specifically detects DGK-delta in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen DGK-delta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 130 kDa diacylglycerol kinase, DAG kinase delta, dgkd-2, DGKdelta, DGK-delta, diacylglycerol kinase delta, diacylglycerol kinase, delta (130kD), diacylglycerol kinase, delta 130kDa, Diglyceride kinase delta, FLJ26930, KIAA0145EC 2.7.1.107
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 960-1140 of human DGKD (NP_690618.2). LPRPPSCSLHPEMLSEEEATQMDQFGQAAGVLIHSIREIAQSHRDMEQELAHAVNASSKSMDRVYGKPRTTEGLNCSFVLEMVNNFRALRSETELLLSGKMALQLDPPQKEQLGSALAEMDRQLRRLADTPWLCQSAEPGDEESVMLDLAKRSRSGKFRLVTKFKKEKNNKNKEAHSSLGA
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8527
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.