Learn More
Delta-Tocopherol, MedChemExpress
Product Code. 16451027
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
16451027 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 16451027 Supplier Medchem Express Supplier No. HY113026
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Delta-Tocopherol is an isomer of Vitamin E.
Delta-Tocopherol is an isomer of Vitamin E.
Specifications
Chemical Name or Material Delta-Tocopherol
CAS 119-13-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H46O2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (124.18 mM; Need ultrasonic)
SMILES OC1=CC(C)=C2C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C1
Molecular Weight (g/mol) 402.65
Formula Weight 402.65
Percent Purity 93.07%
Grade Research
Physical Form Viscous Liquid
For Use With (Application) Metabolism-protein/nucleotide metabolism