Learn More
Dantrolene sodium salt, Thermo Scientific™
Inhibits intracellular calcium release from the sarcoplasmic reticulum
73.80€
Chemical Identifiers
| CAS | 14663-23-1 |
|---|---|
| Fórmula molecular | C14H12N4NaO5 |
| Peso molecular (g/mol) | 339.263 |
| Número MDL | MFCD00079130 |
| Clave InChI | XHONPQLVCHTEDY-YVCISUPJSA-N |
| PubChem CID | 131674048 |
| Nombre IUPAC | molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium |
| SMILES | [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na] |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|
15463579
|
Thermo Scientific Alfa Aesar
J60887.MC |
100mg |
73.80€
100 miligramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 14663-23-1 | |
| 339.263 | |
| XHONPQLVCHTEDY-YVCISUPJSA-N | |
| molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium |
| C14H12N4NaO5 | |
| MFCD00079130 | |
| 131674048 | |
| [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na] |
Specifications
| Dantrolene sodium salt | |
| C14H12N4NaO5 | |
| 14,2816 | |
| [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na] | |
| 339.263 | |
| 336.23 |
| 14663-23-1 | |
| MFCD00079130 | |
| XHONPQLVCHTEDY-YVCISUPJSA-N | |
| molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium | |
| 131674048 |
Safety and Handling
missing translation for 'einecsNumber' : 238-706-8
missing translation for 'rtecsNumber' : MU3875000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures