missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetyl chloride, 97%

Product Code. 11303596
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303596 25 g 25g
11393586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303596 Supplier Thermo Scientific Alfa Aesar Supplier No. L14562.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-99-2 | C7H11ClO | 146.61 g/mol

Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with bases and oxidizing agents. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-99-2
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00236031
InChI Key NILLIUYSJFTTRH-UHFFFAOYSA-N
Synonym cyclopentylacetyl chloride, cyclopentaneacetyl chloride, cyclopentylacetylchloride, cylopentylacetyl chloride, acmc-1brgh, cyclopentaneacetylchloride, cyclopentylacetic chloride, cyclopentanacetyl chloride, cyclopentyl acetyl chloride, cyclopentyl-acetyl chloride
PubChem CID 3485182
IUPAC Name 2-cyclopentylacetyl chloride
SMILES ClC(=O)CC1CCCC1

Specifications

Density 1.087
Boiling Point 186°C
Flash Point 71°C (160°F)
Odor Pungent
Refractive Index 1.4625
Quantity 25 g
UN Number UN3265
Beilstein 1906113
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.62
Percent Purity 97%
Chemical Name or Material Cyclopentylacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.