missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxylic acid, 98%

Product Code. 11467696
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
Product Code. Quantity unitSize
11467696 1000 g 1000g
11477696 250 g 250g
11344539 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11467696

Brand: Thermo Scientific Alfa Aesar A14693.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-89-5 | C7H12O2 | 128.171 g/mol

Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

Solubility
Soluble in water 0.201 g/100 mL (15°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-89-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00001461
InChI Key NZNMSOFKMUBTKW-UHFFFAOYSA-N
Synonym hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
PubChem CID 7413
ChEBI CHEBI:36096
IUPAC Name cyclohexanecarboxylic acid
SMILES C1CCC(CC1)C(=O)O

Specifications

Melting Point 29°C to 32°C
Boiling Point 231°C to 233°C
Flash Point 118°C (244°F)
Quantity 1000 g
Beilstein 970529
Merck Index 14,2724
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Cyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.