Learn More
Sperian™ Honeywell™ Cryogenic Gloves
Product Code. 12201448
Change view
Click to view available options
Size:
9
10
11
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12201448 9 Each
11876920 11 Each
12201458 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12201448 Supplier Sperian™ Supplier No. 205868509
Quantity Price
1 133.32€ / paquete de 1
10 109.29€ / paquete de 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Provides protection against small drops of liquid gas up to -170°C. Sperian™ Honeywell™ Cryogenic Gloves are fully lined with 2 aluminum/fleece layers.
  • Water-repellant silicone cowhide grain leather
  • Gunn cut, winged thumb, crispin style
  • Reinforcement between thumb and index finger
  • 200mm split leather cuff
  • Velcro™ adjustment strap on side of the palm
  • Cuff offers good forearm protection against mechanical hazards and limits any risk of intrusion
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryogenic Gloves
Size 9
Length (Metric) 400 mm