Learn More
Corning™ Universal Fit Pipet Tips: Bulk Packed
GREENER_CHOICE
Product Code. 10163761 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Tip Style:
Beveled, Round
Round
Packaging:
1000 Tips/Bag, 1 Bag/Cs.
1000 Tips/Bag, 10 Bags/Cs.
1000 Tips/Bag, 10,000 Tips/Cs.
1000 Tips/Bag, 1000 Tips/Cs.
Unit Size:
Pack of 1000
Pack of 10000
Product Code. Volume (Metric) Color Tip Style Packaging unitSize
10163761 100 to 1000 μL Blue Round 1000 Tips/Bag, 1 Bag/Cs. Pack of 1000
10380331 1 to 200 μL Natural Beveled, Round 1000 Tips/Bag, 10,000 Tips/Cs. Pack of 10000
10299141 1 to 200 μL Yellow Round 1000 Tips/Bag, 10,000 Tips/Cs. Pack of 10000
10669321 100 to 1000 μL Blue Round 1000 Tips/Bag, 10 Bags/Cs. Pack of 10000
10005451 1 to 200 μL Natural Beveled, Round 1000 Tips/Bag, 1000 Tips/Cs. Pack of 1000
10153761 1 to 200 μL Yellow Round 1000 Tips/Bag, 1000 Tips/Cs. Pack of 1000
6 options
This item is not returnable. View return policy
Product Code. 10163761
Brand: Corning™ 4868
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Designed to assure a reliable fit with major brand pipetters
  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Blue
Format Bulk
Tip Style Round
Packaging 1000 Tips/Bag, 1 Bag/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene