Learn More
STEMCELL Technologies ClonaCell™-HY Hybridoma Kit
Product Code. 17118101
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
17118101 1 Kit 1 set
1 options
This item is not returnable. View return policy
Product Code. 17118101 Supplier STEMCELL Technologies Supplier No. 03800
 Restricted Item
Fecha estimada de envoi: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Media and reagents for all steps of monoclonal antibody generation
  • ClonaCell™-HY Hybridoma Kit contains media and reagents for all steps in hybridoma development and monoclonal antibody production
  • The ClonaCell™-HY method uses a methylcellulose-based semi-solid selective medium to combine hybridoma selection and cloning into one step
  • Individual parental hybridoma clones and their progeny remain localized together in the semi-solid medium as they grow to form distinct colonies
  • This prevents the loss of rare clones by overgrowth from faster-growing cells, as can occur during selection in a liquid medium
  • The hybridoma colonies can be easily picked from the semi-solid medium by manual or robotic methods and dispersed into a liquid growth medium for screening and expansion
  • This kit has been verified for use in mouse and rat hydridoma development and monoclonal antibody production and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster
Specifications
Cell Type Hybridomas
For Use With (Application) Cell Culture; Hybridoma Generation
Quantity 1 Kit