Learn More
Abeba™ Classic 1011 Shoes
Product Code. 15606597
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. Size unitSize
15606597 42 1 pair
15646587 36 1 pair
15656587 37 1 pair
15666587 38 1 pair
15676587 39 1 pair
15686587 40 1 pair
15696587 41 1 pair
15616597 43 1 pair
15626597 44 1 pair
15636597 45 1 pair
15646597 46 1 pair
11 options
This item is not returnable. View return policy
Product Code. 15606597
Brand: Abeba™ 1011/42
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Animal fats, oil and fuel resistant. Leather/microfiber construction.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color Black
Liner Material Microfiber
Material Microfiber
Size 42
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1011
Toe Material Steel
Resistant To Animal fats, oil and fuel