missing translation for 'onlineSavingsMsg'
Learn More

Chlorodi(1-naphthyl)phosphine, 95%

Product Code. 11485640
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11485640 5 g 5g
11475640 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11485640

Brand: Thermo Scientific Alfa Aesar H25893.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 36042-99-6 | C20H14ClP | 320.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36042-99-6
Molecular Formula C20H14ClP
Molecular Weight (g/mol) 320.76
MDL Number MFCD09753469
InChI Key ZHRMSAAMUUNWKE-UHFFFAOYSA-N
Synonym chlorodi 1-naphthyl phosphine, bis 1-naphthyl chlorophosphine, phosphinous chloride, di-1-naphthalenyl, chlorobis naphthalen-1-yl phosphane, di 1-naphthyl chlorophosphine, chloro dinaphthalen-1-yl phosphane, dinaphthalen-1-ylphosphinous chloride, phosphinous chloride,p,p-di-1-naphthalenyl
PubChem CID 18705941
SMILES ClP(C1=CC=CC2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 154°C to 157°C
Boiling Point 227°C to 229°C (0.6 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name chlorobis(naphthalen-1-yl)phosphane
Formula Weight 320.76
Percent Purity 95%
Chemical Name or Material Chlorodi(1-naphthyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.