Learn More
Globe Scientific Certified Microcentrifuge Tubes in Self-Standing Bags
Descripción
- Manufactured from FDA approved resins
- Certified RNase, DNase, DNA and Pyrogen free
- Supplied in resealable, tamper-evident self-standing bags
- Large, clearly printed graduations
- Frosted writing area on each tube
- Smooth surfaces won't catch or tear gloves
- Thin cap membrane for trouble-free needle piercing
- Fit all standard and high capacity centrifuge rotors
- Can be centrifuged at up to 20,000 RCF
- Withstand temperatures from −80°C to +121°C
- Food grade
Especificaciones
Especificaciones
| Certificaciones/conformidad | RNase, DNase, DNA and Pyrogen Free |
| Esterilizables en autoclave | Autoclavable |
| Capacidad (métrico) | 2.0 mL |
| Color | Natural |
| Material | Polypropylene |
| N.° por paquete | 500 |
| Envase | Self-standing tamper evident bag |
| Volumen (métrico) | 2mL |
| Esterilidad | Non-sterile |
| Graduated | Yes |
| Mostrar más |