Learn More
Unity Lab Services™ Centrifuge Plus Requalification Service
Product Code. 30178676
Change view
Click to view available options
For Use With (Equipment):
Centrifuge
Unit Size:
1 unit
Product Code. For Use With (Equipment) Unit Size
30178676 Centrifuge 1 unit
1 options
This item is not returnable. View return policy
Product Code. 30178676 Supplier Unity Lab Services™ Supplier No. RQCENPLUS
 Restricted Item
Fecha estimada de envoi: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Requalification Services for Centrifuge Plus (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests Min/Max speed, Brief /Extended time, temperature for refrigerated models.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Centrifuge
Product Type Requalification Service