Learn More
Cefmetazole sodium, 921μg/mg, Thermo Scientific™
120.00€
| CAS | 56796-39-5 |
|---|---|
| Fórmula molecular | C15H16N7NaO5S3 |
| Peso molecular (g/mol) | 493.507 |
| Número MDL | MFCD00214260 |
| Clave InChI | BITQGIOJQWZUPL-PBCQUBLHSA-M |
| PubChem CID | 23666711 |
| ChEBI | CHEBI:3490 |
| Nombre IUPAC | sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| SMILES | CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+] |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15447669
|
Thermo Scientific Alfa Aesar
J66348.03 |
1g |
120.00€
1 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis
- It is also used to study protein mediated antibiotic tr
| 56796-39-5 | |
| 493.507 | |
| BITQGIOJQWZUPL-PBCQUBLHSA-M | |
| CHEBI:3490 | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+] |
| C15H16N7NaO5S3 | |
| MFCD00214260 | |
| 23666711 | |
| sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Specifications
| Cefmetazole sodium | |
| 56796-39-5 | |
| Hygroscopic | |
| MFCD00214260 | |
| BITQGIOJQWZUPL-PBCQUBLHSA-M | |
| sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 23666711 | |
| 493.52 |
| 921μg/mg | |
| White | |
| C15H16N7NaO5S3 | |
| Soluble in water | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+] | |
| 493.507 | |
| CHEBI:3490 |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
missing translation for 'rtecsNumber' : XI0372200
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen