Learn More
SI Analytics™ Cannon-Fenske Reverse Flow Viscometer
Product Code. 10423711 Shop All SI Analytics Products
Change view
Click to view available options
Viscosity Range:
20 to 100 cSt
3 to 15 cSt
7 to 35 cSt
Approximate Constant:
0.0215K
0.035K
0.1K
Size:
0.63mm Diameter
0.78mm Diameter
1.02mm Diameter
Unit Size:
Each
Product Code. Viscosity Range Approximate Constant Size unitSize
10423711 20 to 100 cSt 0.1K 1.02mm Diameter Each
10707021 3 to 15 cSt 0.0215K 0.63mm Diameter Each
10538981 7 to 35 cSt 0.035K 0.78mm Diameter Each
3 options
This item is not returnable. View return policy
Product Code. 10423711
Brand: SI Analytics™ 285403059
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Cannon-Fenske Reverse Flow Viscometer includes three ring marks since they can only be checked by means of manual measurements.
  • Suitable for transparent liquids manual measurement, opaque liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • Calibrated
  • With two constants
  • With three ring marks
  • Only for manual measurements
  • Filling quantity of 12mL
  • Overall length of 295mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 20 to 100 cSt
Calibrated Yes
Description Capillary No. 200
Approximate Constant 0.1K
ASTM Numbers D 446
Certifications/Compliance ISO/DIS 3105, ASTMD 2515, ASTM D 446, NF T 60 - 100
Includes Ring Marks and 2 Constants
For Use With (Application) Only for manual measurement
Size 1.02mm Diameter
Volume (Metric) 12 mL