Learn More
Calibration Buffer Solutions, Hanna Instruments™
Click to view available options
Quantity:
20 mL
230 mL
1 L
pH:
4.01
9.18
10.01
Unit Size:
Each
Pack of 25
Description
- A premium quality pH 4.01 calibration solution that has been dyed red for easy buffer identification
- pH 4.01 calibration buffer produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Specifications
| Color | Colorless |
| pH | 4.01 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
| Formulation | Aqueous Buffer Solution |
| Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction