missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%

Product Code. 11486190
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486190 5 g 5g
11476190 1 g 1g
11496190 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486190 Supplier Thermo Scientific Alfa Aesar Supplier No. H27106.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56210-72-1 | C16H19N | 225.335 g/mol

(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56210-72-1
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.335
MDL Number MFCD00243087
InChI Key NXLACVVNHYIYJN-KBPBESRZSA-N
Synonym --bis s-1-phenylethyl amine, s-bis s-1-phenylethyl amine, unii-5d97t2y7wu, bis s-1-phenylethyl amine, 1s-1-phenyl-n-1s-1-phenylethyl ethanamine, bis 1s-1-phenylethyl amine, s,s-di 1-phenylethyl amine
PubChem CID 6994958
IUPAC Name (1S)-1-phenyl-N-[(1S)-1-phenylethyl]ethanamine
SMILES CC(C1=CC=CC=C1)NC(C)C2=CC=CC=C2

Specifications

Boiling Point 98°C (0.3 mmHg)
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 225.34
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (-)-Bis[(S)-1-phenylethyl]amine
compliance-icons
Product Identifier
  • (-)-Bis[(S)-1-phenylethyl]amine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (-)-Bis[(S)-1-phenylethyl]amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.