Learn More
Bis(phenylthio)methane, 97%, BLD Pharm™
Product Code. 30968241
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968241 100 g 100g
30966898 5 g 5g
30951322 1 g 1g
30958418 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30968241 Supplier BLD Pharm Supplier No. BD117218100g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3561-67-9
Molecular Formula C13H12S2
MDL Number MFCD00003067
Quantity 100 g
SMILES C1(SCSC2=CC=CC=C2)=CC=CC=C1
Molecular Weight (g/mol) 232.36
Percent Purity 97%
Chemical Name or Material Bis(phenylthio)methane