Learn More
Benzyl 2-chloroacetate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 140-18-1 |
| Molecular Formula | C9H9ClO2 |
| MDL Number | MFCD00157062 |
| Quantity | 500 g |
| SMILES | O=C(OCC1=CC=CC=C1)CCl |
| Molecular Weight (g/mol) | 184.62 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzyl 2-chloroacetate |