Learn More
BD Human IL-1β Set II
Product Code. 15859128 Shop All BD Biosciences Products
Change view
Click to view available options
Concentration:
3.9 to 250pg/mL
Unit Size:
Each
Product Code. Concentration Unit Size
15859128 3.9 to 250pg/mL Each
1 options
This item is not returnable. View return policy
Product Code. 15859128 Supplier BD Supplier No. 557953
 Restricted Item
Fecha estimada de envoi: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Includes
Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
An immunoassay design to fit individual laboratory needs
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IL-1β in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Concentration 3.9 to 250pg/mL
For Use With (Application) ELISA
Name Human IL-1 beta Set II
Quantity 20 Plates
Storage Requirements -80°C
Cross Reactivity Human