missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: m LES Endo Agar
Description
- Formulation: Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g
Certifications
CE, COMPF, SMD, SMWW
Specifications
Specifications
| Certifications/Compliance | CE, COMPF, SMD, SMWW |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | mLES Endo Agar |
| Product Type | Agar |
| Formulation | Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g/L, Basic Fuchsin 0.8g/L, Agar 15g/L |
| For Use With (Application) | Coliform enumeration |
| Packaging Type | Bottle |
| Quantity | 100 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?