Specifications
| Antigen | Avidin D |
| Quantity | 100 Units |
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Line | Novus |
| For Use With (Application) | To avoid adsorption or denaturing of the enzyme conjugate, dilutions may be made in 20 mM HEPES, 0.5 M NaCl, pH 7.5, containing 0.1 mg/mL IHC- grade BSA. DO NOT USE FRACTION V GRADE BSA. One unit equals 1.0 micromole of p-nitrophenol liberated from p-nitrophenylphosphate per minute at 37C (pH 9.5). Recommended concentration for use is 0.2 units/mL. Storage of dilute solutions of the enzyme conjugate is not recommended. |